Learn More
Biomatik Corporation Rat Platelet-Derived Growth Factor-Bb, Pdgf-Bb Elisa Kit
Supplier:  Biomatik Corporation EKC39566
Encompass_Preferred
Rat Platelet-Derived Growth Factor-Bb, Pdgf-Bb Elisa Kit for detecting Platelet-Derived Growth Factor-Bb, Pdgf-Bb in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 39.063-2500pg/ml. Sensitivity: 9.77pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4629