Learn More
Biomatik Corporation Rat Protein S100-A1(S100A1) ELISA kit
Supplier:  Biomatik Corporation EKC39608
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35467. Target Synonyms: S100, S100-alpha, S100A, S100 alpha|S100 calcium-binding protein A1|S100 protein, alpha polypeptide. Restriction: For Research Use Only.
Catalog No. 50-229-4631