Learn More
Biomatik Corporation Rat Thioredoxin,Trx ELISA Kit
Supplier:  Biomatik Corporation EKC39759
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P11232. Target Synonyms: RP11-427L11.1, DKFZp686B1993, MGC61975, TRX, TRX1,. Restriction: For Research Use Only.
Catalog No. 50-229-4650