Learn More
Biomatik Corporation Rat Tumor necrosis factor soluble receptor Ⅰ,TNFsR-Ⅰ ELISA KIT
Supplier:  Biomatik Corporation EKC39836
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P22934. Target Synonyms: CD120a, FPF, MGC19588, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR1, TNFR55, TNFR60, p55, p55-R, p60, tumor necrosis factor binding protein 1|tumor necrosis factor receptor 1, TNFsR-1. Restriction: For Research Use Only.
Catalog No. 50-229-4659