Learn More
Biomatik Corporation Rat vimentin (VIM)ELISA Kit
Supplier:  Biomatik Corporation EKC39874
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 23.5 ng/mL-1500 ng/mL. Sensitivity: 5.8 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P31000. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4664