Learn More
Biomatik Corporation Rhesus thrombin-antithrombin complex (TAT)ELISA Kit
Supplier:  Biomatik Corporation EKC39903
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Rhesus. Detection Range: 1.25 pg/mL-80 pg/mL. Sensitivity: 0.312 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4666