Learn More
Biomatik Corporation Human Erythropoietin Receptor, Epor Elisa Kit
Supplier:  Biomatik Corporation EKC40037
Encompass_Preferred
Human Erythropoietin Receptor, Epor Elisa Kit for detecting Erythropoietin Receptor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19235; EPOR. Target Synonyms: EPOR; Erythropoietin receptor; EPO-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4681