Learn More
Biomatik Corporation Human Factor-Related Apoptosis, Fas Elisa Kit
Supplier:  Biomatik Corporation EKC40039
Encompass_Preferred
Human Factor-Related Apoptosis, Fas Elisa Kit for detecting Fas (Tnf Receptor Superfamily, Member 6) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P25445; FAS. Target Synonyms: FAS; APT1; FAS1; TNFRSF6; Tumor necrosis factor receptor superfamily member 6; Apo-1 antigen; Apoptosis-mediating surface antigen FAS; FASLG receptor; CD antigen CD95. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4683