Learn More
Biomatik Corporation Mouse Fractalkine, Fk Elisa Kit
Supplier:  Biomatik Corporation EKC40046
Encompass_Preferred
Mouse Fractalkine, Fk Elisa Kit for detecting Chemokine (C-X3-C Motif) Ligand 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35188; Cx3cl1. Target Synonyms: Cx3cl1; Cx3c; Fkn; Scyd1; Fractalkine; FK; C-X3-C motif chemokine 1; CX3C membrane-anchored chemokine; Neurotactin; Small-inducible cytokine D1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4685