Learn More
Biomatik Corporation Mouse Insulin-like growth factor binding protein 1,IGFBP-1 ELISA Kit
Supplier:  Biomatik Corporation EKC40061
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P47876. Target Synonyms: AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1, IGF-binding protein 1|alpha-pregnancy-associated endometrial globulin|amniotic fluid binding protein|binding protein-25|binding protein-26|binding protein-28|gr. Restriction: For Research Use Only.
Catalog No. 50-229-4689