Learn More
Biomatik Corporation Human soluble interleukin-1 receptor Ⅰ,IL-1sRⅠ ELISA kit
Supplier:  Biomatik Corporation EKC40074
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/ml-10 ng/ml. Sensitivity: 0.039 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P14778. Target Synonyms: CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80, OTTHUMP00000161344|antigen CD121a|interleukin 1 receptor alpha, type I|interleukin receptor 1. Restriction: For Research Use Only.
Catalog No. 50-229-4694