Learn More
Biomatik Corporation Human Urokinase-Type Plasminogen Activator Receptor, Plaur/Upar Elisa Kit
Supplier:  Biomatik Corporation EKC40126
Encompass_Preferred
Human Urokinase-Type Plasminogen Activator Receptor, Plaur/Upar Elisa Kit for detecting Urokinase-Type Plasminogen Activator Receptor, Plaur/Upar in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q03405; PLAUR. Target Synonyms: CD 87; CD87; CD87 antigen; MO 3; MO3; Monocyte activation antigen Mo3; Plasminogen activator receptor urokinase; Plasminogen activator urokinase receptor; PLAUR; U PAR; u plasminogen activator receptor; U-PAR; u-plasminogen activator receptor form 2; UPA receptor; uPAR; UPAR_HUMAN; Urinary plasminogen activator receptor; URKR; Urokinase plasminogen activator receptor; Urokinase plasminogen activator surface receptor; urokinase-type plasminogen activator (uPA) receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4703