Learn More
Biomatik Corporation Human Vascuar Endothelial Cell Growth Factor Receptor 3, Vegfr-3/Flt-4 Elisa Kit
Supplier:  Biomatik Corporation EKC40132
Encompass_Preferred
Human Vascuar Endothelial Cell Growth Factor Receptor 3, Vegfr-3/Flt-4 Elisa Kit for detecting Fms-Related Tyrosine Kinase 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35916; FLT4. Target Synonyms: EC 2.7.10.1; flt 4; FLT-4; FLT4; FLT41; Fms related tyrosine kinase 4; Fms-like tyrosine kinase 4; LMPH1A; PCL; Soluble VEGFR3 variant 1; Soluble VEGFR3 variant 2; Soluble VEGFR3 variant 3; Tyrosine protein kinase receptor FLT4; Tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; Vascular endothelial growth factor receptor 3 precursor; VEGF R3; VEGFR 3; VEGFR-3; VEGFR3; VGFR3_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4706