Learn More
Biomatik Corporation Human Myoglobin, Myo/Mb Elisa Kit
Supplier:  Biomatik Corporation EKC40205
Encompass_Preferred
Human Myoglobin, Myo/Mb Elisa Kit for detecting Myoglobin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02144; MB. Target Synonyms: MB; MGC13548; MYG_HUMAN; Myoglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4713