Learn More
Biomatik Corporation Rat Insulin-like growth factor binding protein 3,IGFBP-3 ELISA kit
Supplier:  Biomatik Corporation EKC40303
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 2.7 ng/ml - 2000 ng/ml. Sensitivity: 2.7 ng/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15473. Target Synonyms: tcag7.703, BP-53, IBP3, IGF-binding protein 3|acid stable subunit of the 140 K IGF complex|binding protein 29|binding protein 53|growth hormone-dependent binding protein. Restriction: For Research Use Only.
Catalog No. 50-229-4726