Learn More
Biomatik Corporation Rat Interleukin 11,IL-11 ELISA KIT
Supplier:  Biomatik Corporation EKC40305
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q99MF5. Target Synonyms: AGIF, IL-11, adipogenesis inhibitory factor|oprelvekin. Restriction: For Research Use Only.
Catalog No. 50-229-4728