Learn More
Biomatik Corporation Rat Endostatin,ES ELISA Kit
Supplier:  Biomatik Corporation EKC40360
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: FLJ27325, FLJ34914, KNO, KNO1, MGC74745, alpha 1 type XVIII collagen|antiangiogenic agent|endostatin|multi-functional protein MFP. Restriction: For Research Use Only.
Catalog No. 50-229-4746