Learn More
Biomatik Corporation Mouse Cholecystokinin,CCK ELISA Kit
Supplier:  Biomatik Corporation EKC40385
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09240. Target Synonyms: MGC117187, OTTHUMP00000209369. Restriction: For Research Use Only.
Catalog No. 50-229-4750