Learn More
Biomatik Corporation Rat Thyroid-Peroxidase,TPO ELISA Kit
Supplier:  Biomatik Corporation EKC40430
Encompass_Preferred
Sample Type: serum, plasma, tissue Homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 2.5 ng/mL-40ng/mL. Sensitivity: 2.5 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P14650. Target Synonyms: MSA, TDH2A, TPX, OTTHUMP00000115536|thyroid microsomal antigen|thyroperoxidase. Restriction: For Research Use Only.
Catalog No. 50-229-4753