Learn More
Biomatik Corporation Human Osteonectin, On Elisa Kit
Supplier:  Biomatik Corporation EKC40512
Encompass_Preferred
Human Osteonectin, On Elisa Kit for detecting Sparc/Osteonectin, Cwcv And Kazal-Like Domains Proteoglycan (Testican) 2 in serum, plasma, saliva, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92563; SPOCK2. Target Synonyms: and Kazal-like domains proteoglycan 2; CWCV; FLJ97039; KIAA0275; SPARC/osteonectin; Sparc/osteonectin cwcv and kazal like domains proteoglycan (testican) 2; Sparc/osteonectin cwcv and kazal like domains proteoglycan 2; SPOCK 2; Spock2; Testican 2; Testican-2; Testican2; TICN 2; TICN2; TICN2_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4760