Learn More
Biomatik Corporation Human Carbonic Anhydrase 2, Ca-2 Elisa Kit
Supplier:  Biomatik Corporation EKC40524
Encompass_Preferred
Human Carbonic Anhydrase 2, Ca-2 Elisa Kit for detecting Carbonic Anhydrase Ii in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00918; CA2. Target Synonyms: CA 2; CA II; CA-II; Ca2; CAC; CAH2_HUMAN; CAII; Car 2; Car2; Carbonate dehydratase II; Carbonic anhydrase 2; Carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase C; formerly; Carbonic anhydrase II; Carbonic dehydratase; epididymis luminal protein 76; Epididymis secretory protein Li 282; HEL-76; HEL-S-282. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4762