Learn More
Biomatik Corporation Mouse Caspase 3,Casp-3 ELISA Kit
Supplier:  Biomatik Corporation EKC40529
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P70677. Target Synonyms: CPP32, CPP32B, SCA-1, PARP cleavage protease|SREBP cleavage activity 1|Yama|apopain|caspase 3|caspase 3, apoptosis-related cysteine protease|cysteine protease CPP32|procaspase3. Restriction: For Research Use Only.
Catalog No. 50-229-4764