Learn More
Biomatik Corporation Rat Cathepsin K,cath-K ELISA Kit
Supplier:  Biomatik Corporation EKC40623
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O35186. Target Synonyms: RP11-363I22.4, CTS02, CTSO, CTSO1, CTSO2, MGC23107, PKND, PYCD, cathepsin O1|cathepsin O2|cathepsin X. Restriction: For Research Use Only.
Catalog No. 50-229-4774