Learn More
Biomatik Corporation Human Heparin-Binding Epidermal Growth Factor-Like Growth Factor, Hb-Egf Elisa Kit
Supplier:  Biomatik Corporation EKC40676
Encompass_Preferred
Human Heparin-Binding Epidermal Growth Factor-Like Growth Factor, Hb-Egf Elisa Kit for detecting Heparin-Binding Egf-Like Growth Factor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99075; HBEGF. Target Synonyms: diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); Diphtheria toxin receptor; DT R; DT-R; DTR; DTS; DTSF; HB-EGF; HBEGF; HBEGF_HUMAN; HEGFL; Heparin binding EGF like growth factor; Heparin binding epidermal growth factor; Heparin binding epidermal growth factor like growth factor; Heparin-binding EGF-like growth factor; Proheparin binding EGF like growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4776