Learn More
Biomatik Corporation Monkey monocyte chemotactic protein 1/monocyte chemotactic and activating factor,MCP1/MCAF ELISA kit
Supplier:  Biomatik Corporation EKC40733
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P61275. Target Synonyms: GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, MGC9434, SCYA2, SMC-CF, monocyte chemoattractant protein-1|monocyte chemotactic and activating factor|monocyte secretory protein JE|small inducible cytokine. Restriction: For Research Use Only.
Catalog No. 50-229-4788