Learn More
Biomatik Corporation Rat apolipoprotein H (Apo-H) ELISA Kit
Supplier:  Biomatik Corporation EKC40741
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 28 ng/mL-1800 ng/mL. Sensitivity: 7 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P26644. Target Synonyms: B2G1, BG, apolipoprotein H. Restriction: For Research Use Only.
Catalog No. 50-229-4790