Learn More
Biomatik Corporation Human Prostaglandin F2Α, Pgf2Α Elisa Kit
Supplier:  Biomatik Corporation EKC40755
Encompass_Preferred
Human Prostaglandin F2Α, Pgf2Α Elisa Kit for detecting Prostaglandin F2Α, Pgf2Α in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: PGF2 alpha; Prostaglandin F2 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4792