Learn More
Biomatik Corporation Human Insulin-Like Growth Factors 2 Receptor, Igf-2R Elisa Kit
Supplier:  Biomatik Corporation EKC40792
Encompass_Preferred
Human Insulin-Like Growth Factors 2 Receptor, Igf-2R Elisa Kit for detecting Insulin-Like Growth Factor 2 Receptor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 312.5-20000pg/ml. Sensitivity: 78.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11717; IGF2R. Target Synonyms: 300 kDa mannose 6 phosphate receptor; 300 kDa mannose 6-phosphate receptor; Cation independent mannose 6 phosphate receptor; Cation-independent mannose-6-phosphate receptor; CD222; CD222 antigen; CI Man 6 P receptor; CI Man-6-P receptor; CI MPR; CI-M6PR; CI-MPR; CIMPR; IGF 2 receptor; IGF 2R; IGF II receptor; IGF-II receptor; IGF2 receptor; Igf2r; Insulin like growth factor 2 receptor; Insulin like growth factor II receptor; Insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; M6P R; M6P/IGF2 receptor; M6P/IGF2R; M6PR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4797