Learn More
Biomatik Corporation Human collectin sub-family member 11,COLEC11 ELISA Kit
Supplier:  Biomatik Corporation EKC40819
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2 pg/mL-2000 pg/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BWP8. Target Synonyms: CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, CLK1, DKFZp686N1868, MGC3279, collectin kidney I. Restriction: For Research Use Only.
Catalog No. 50-229-4800