Learn More
Biomatik Corporation Mouse Myeloid Differentiation Factor 88, Myd88 Elisa Kit
Supplier:  Biomatik Corporation EKC40848
Encompass_Preferred
Mouse Myeloid Differentiation Factor 88, Myd88 Elisa Kit for detecting Myeloid Differentiation Primary Response Gene (88) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 47-3000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22366; Myd88. Target Synonyms: Myd88; Myeloid differentiation primary response protein MyD88. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4805