Learn More
Biomatik Corporation Human Insulin-Like Growth Factor Binding Protein 5, Igfbp-5 Elisa Kit
Supplier:  Biomatik Corporation EKC40965
Encompass_Preferred
Human Insulin-Like Growth Factor Binding Protein 5, Igfbp-5 Elisa Kit for detecting Insulin-Like Growth Factor Binding Protein 5 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P24593; IGFBP5. Target Synonyms: IBP 5; IBP-5; IBP5; IBP5_HUMAN; IGF binding protein 5; IGF BP5; IGF-binding protein 5; IGFBP 5; IGFBP-5; IGFBP5; Insulin like growth factor binding protein 5; Insulin-like growth factor-binding protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4811