Learn More
Biomatik Corporation Human Interleukin 28B (Il-28B) Elisa Kit
Supplier:  Biomatik Corporation EKC40973
Encompass_Preferred
Human Interleukin 28B (Il-28B) Elisa Kit for detecting Interleukin 28B(Il-28B) in serum, plasma, cell culture supernats, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8IZI9; IFNL3. Target Synonyms: Cytokine ZCYTO22; IFN-lambda-3; IFN-lambda-4; IL-28B; IL-28C; IL28B; IL28B_HUMAN; IL28C; Interferon lambda-3; Interferon lambda-4; interleukin 28B (interferon; lambda 3); Interleukin 28C; Interleukin-28B; Interleukin-28C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4814