Learn More
Biomatik Corporation Human left-right determination factor 2(LEFTY2) ELISA Kit
Supplier:  Biomatik Corporation EKC41031
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O00292. Target Synonyms: EBAF, LEFTA, LEFTYA, MGC46222, TGFB4, endometrial bleeding associated factor|endometrial bleeding associated factor (left-right determination, factor A; transforming growth factor beta superfamily)|. Restriction: For Research Use Only.
Catalog No. 50-229-4823