Learn More
Biomatik Corporation Goat Testosterone (T) ELISA kit
Supplier:  Biomatik Corporation EKC41040
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Capra hircus (Goat). Detection Range: 0.1 ng/mL-20 ng/mL. Sensitivity: 0.05 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4827