Learn More
Biomatik Corporation Dog Albumin (Alb) Elisa Kit
Supplier:  Biomatik Corporation EKC41172
Encompass_Preferred
Dog Albumin (Alb) Elisa Kit for detecting Albumin in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.273-35ug/ml. Sensitivity: 0.266ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P49822; ALB. Target Synonyms: ALBSerum albumin; allergen Can f 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4847