Learn More
Biomatik Corporation Human Cystatin-B (Cstb/Cst6/Stfb) Elisa Kit
Supplier:  Biomatik Corporation EKC41239
Encompass_Preferred
Human Cystatin-B (Cstb/Cst6/Stfb) Elisa Kit for detecting Cystatin B (Stefin B) in serum, plasma, cell culture supernates, urine, saliva, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.233ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04080; CSTB. Target Synonyms: CHROW21; CPI B; CPI-B; CST 6; CST6; CSTB; Cystatin B (stefin B); Cystatin B; Cystatin-B; CYTB; CYTB_HUMAN; EPM1; EPM1A; Liver thiol proteinase inhibitor; PME; Stefin-B; STF B; STFB; ULD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4855