Learn More
Biomatik Corporation Human Interleukin 19 (Il-19) Elisa Kit
Supplier:  Biomatik Corporation EKC41278
Encompass_Preferred
Human Interleukin 19 (Il-19) Elisa Kit for detecting Interleukin 19 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UHD0; IL19. Target Synonyms: IL 10C; IL 19; IL-19; Il19; IL19_HUMAN; Interleukin 19; Interleukin-19; MDA1; Melanoma differentiation-associated protein-like protein; NG.1; ZMDA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4857