Learn More
Biomatik Corporation Rat selenoprotein P, plasma, 1 (SEPP1)ELISA kit
Supplier:  Biomatik Corporation EKC41308
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P25236. Target Synonyms: SELP, SeP, selenoprotein P. Restriction: For Research Use Only.
Catalog No. 50-229-4860