Learn More
Biomatik Corporation Human Myelin-Associated Glycoprotein (Mag) Elisa Kit
Supplier:  Biomatik Corporation EKC41324
Encompass_Preferred
Human Myelin-Associated Glycoprotein (Mag) Elisa Kit for detecting Myelin Associated Glycoprotein in serum, plasma, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20916; MAG. Target Synonyms: GMA; MAG; MAG_HUMAN; Myelin associated glycoprotein; Myelin-associated glycoprotein; S MAG; S-MAG; Sialic acid binding Ig like lectin 4A; Sialic acid binding immunoglobulin like lectin 4A; Siglec 4a; Siglec-4a; SIGLEC4A; SPG75. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4862