Learn More
Biomatik Corporation Human Apolipoprotein D (Apod) Elisa Kit
Supplier:  Biomatik Corporation EKC41364
Encompass_Preferred
Human Apolipoprotein D (Apod) Elisa Kit for detecting Apolipoprotein D in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05090; APOD. Target Synonyms: APO D; Apo-D; ApoD; APOD protein; APOD_HUMAN; Apolipoprotein D; ApolipoproteinD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4866