Learn More
Biomatik Corporation Human Caspase-7 (Casp7) Elisa Kit
Supplier:  Biomatik Corporation EKC41412
Encompass_Preferred
Human Caspase-7 (Casp7) Elisa Kit for detecting Caspase 7, Apoptosis-Related Cysteine Peptidase in serum, plasma, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55210; CASP7. Target Synonyms: apoptosis-related cysteine peptidase; Apoptotic protease Mch-3; CASP-7; CASP7; CASP7_HUMAN; Caspase-7 subunit p11; CMH-1; ICE-LAP3; ICE-like apoptotic protease 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4870