Learn More
Biomatik Corporation Human B-Cell Differentiation Antigen Cd72 (Cd72) Elisa Kit
Supplier:  Biomatik Corporation EKC41427
Encompass_Preferred
Human B-Cell Differentiation Antigen Cd72 (Cd72) Elisa Kit for detecting Cd72 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21854; CD72. Target Synonyms: CD72; B-cell differentiation antigen CD72; Lyb-2; CD antigen CD72. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4872