Learn More
Biomatik Corporation Human Defensin-6 (Defa6) Elisa Kit
Supplier:  Biomatik Corporation EKC41518
Encompass_Preferred
Human Defensin-6 (Defa6) Elisa Kit for detecting Defensin, Alpha 6, Paneth Cell-Specific in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q01524; DEFA6. Target Synonyms: alpha 6; DEF6; DEF6_HUMAN; DEFA 6; DEFA6; Defensin 6; Defensin alpha 6; Defensin alpha 6 Paneth cell specific; Defensin; Defensin-6; HD 6; HD6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4884