Learn More
Biomatik Corporation Human Gastrotropin (Fabp6) Elisa Kit
Supplier:  Biomatik Corporation EKC41551
Encompass_Preferred
Human Gastrotropin (Fabp6) Elisa Kit for detecting Fatty Acid Binding Protein 6, Ileal in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51161; FABP6. Target Synonyms: Fabp6; FABP6_HUMAN; Fatty acid binding protein 6; ileal (gastrotropin); Fatty acid-binding protein 6; Gastrotropin; GT; I 15P; I BABP; I BALB; I BAP; I-15P; I-BABP; I15P; IBABP; IBALB; IBAP; ILBP; ILBP3; Ileal lipid binding protein; Ileal lipid-binding protein; ILLBP; Intestinal 15 kDa protein; Intestinal bile acid binding protein; Intestinal bile acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4888