Learn More
Biomatik Corporation Human Fetuin-B (Fetub) Elisa Kit
Supplier:  Biomatik Corporation EKC41563
Encompass_Preferred
Human Fetuin-B (Fetub) Elisa Kit for detecting Fetuin B in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UGM5; FETUB. Target Synonyms: 16G2; 2310011O17Rik; AI255764; D17980; Fet; Fetub; FETUB_HUMAN; Fetuin beta; Fetuin-B; Fetuin-like protein; Fetuin-like protein IRL685; Gugu; IRL685; Pp63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4890