Learn More
Biomatik Corporation Bovine Heparin-Binding Growth Factor 2 (Fgf2) Elisa Kit
Supplier:  Biomatik Corporation EKC41570
Encompass_Preferred
Bovine Heparin-Binding Growth Factor 2 (Fgf2) Elisa Kit for detecting Fibroblast Growth Factor 2 (Basic) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P03969; FGF2. Target Synonyms: FGF2Fibroblast growth factor 2; FGF-2; Basic fibroblast growth factor; bFGF; Heparin-binding growth factor 2; HBGF-2) [Cleaved into: Kidney-derived growth factor]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4892