Learn More
Biomatik Corporation Human Fibroblast Growth Factor Receptor 3 (Fgfr3) Elisa Kit
Supplier:  Biomatik Corporation EKC41576
Encompass_Preferred
Human Fibroblast Growth Factor Receptor 3 (Fgfr3) Elisa Kit for detecting Fibroblast Growth Factor Receptor 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22607; FGFR3. Target Synonyms: ACH; CD 333; CD333; CD333 antigen; CEK 2; CEK2; FGFR 3; FGFR-3; FGFR3; FGFR3_HUMAN; Fibroblast growth factor receptor 3 (achondroplasia thanatophoric dwarfism); Fibroblast growth factor receptor 3; Heparin binding growth factor receptor; HSFGFR3EX; Hydroxyaryl protein kinase; JTK 4; JTK4; MFR 3; SAM 3; Tyrosine kinase JTK 4; Tyrosine kinase JTK4; Z FGFR 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4894