Learn More
Biomatik Corporation Human Glypican 1 (Gpc1) Elisa Kit
Supplier:  Biomatik Corporation EKC41606
Encompass_Preferred
Human Glypican 1 (Gpc1) Elisa Kit for detecting Glypican 1 (Gpc1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35052; GPC1. Target Synonyms: FLJ38078; glypican; Glypican proteoglycan 1; glypican1; GPC 1; Gpc1; GPC1_HUMAN; Secreted glypican-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4900