Learn More
Biomatik Corporation Bovine Heat Shock Protein Hsp 90-Alpha (Hsp90Aa1) Elisa Kit
Supplier:  Biomatik Corporation EKC41632
Encompass_Preferred
Bovine Heat Shock Protein Hsp 90-Alpha (Hsp90Aa1) Elisa Kit for detecting Heat Shock Protein 90Kda Alpha (Cytosolic), Class A Member 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Bovine (Bos taurus). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q76LV2; HSP90AA1. Target Synonyms: HSP90AA1; HSP90A; HSPCA; Heat shock protein HSP 90-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4905