Learn More
Biomatik Corporation Human Icos Ligand (Icoslg) Elisa Kit
Supplier:  Biomatik Corporation EKC41643
Encompass_Preferred
Human Icos Ligand (Icoslg) Elisa Kit for detecting Inducible T-Cell Co-Stimulator Ligand in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 23.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75144; ICOSLG. Target Synonyms: ICOSLG; B7H2; B7RP1; ICOSL; KIAA0653; ICOS ligand; B7 homolog 2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP-1; CD antigen CD275. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4908